Description
- Calibration certificate
- For temperature
Specifications
Specifications
| For Use With (Equipment) | BINDER Chambers Series KB ECO |
| Description | Calibration certificate, temperature - for temperature, measurement in center of chamber at specified temperature |
| Duration | 60 to 120 min. |
| Calibration Type | 1 P Temperature |
| Product Type | Calibration Certificate |