Learn More
BINDER™ Calibration Certificate for BINDER™ KB ECO Series
Product Code. 17502875 Shop All BINDER Products
Change view
Click to view available options
Calibration Type:
1 P Expanded Temperature
1 P Temperature
18 P Temperature
27 P Temperature
9 P Temperature
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Calibration Type unitSize
17502875 1 P Temperature Each
17512875 1 P Expanded Temperature Each
17522875 9 P Temperature Each
17532875 18 P Temperature Each
17542875 27 P Temperature Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 17502875 Supplier BINDER™ Supplier No. 80122106
The Calibration Certificate for BINDER KB ECO Series is an option for KB ECO Series Refrigerated Incubators with environmentally friendly Thermoelectric Cooling.
  • Calibration certificate
  • For temperature
Specifications
For Use With (Equipment) BINDER Chambers Series KB ECO
Description Calibration certificate, temperature - for temperature, measurement in center of chamber at specified temperature
Duration 60 to 120 min.
Calibration Type 1 P Temperature
Product Type Calibration Certificate