Learn More
Hanna Instruments™ Calibration Buffer pH 10.01, FDA Bottle
Product Code. 11733193
missing translation for 'orderingAttributeHoverText'
Quantity:
500 mL
missing translation for 'unitSize'
Each
Product Code. 11733193
missing translation for 'mfr': Hanna Instruments™ HI8010L/C
Specifications
Boiling Point ∼100°C
Color Colorless
Composition Aqueous Buffer Solution
Filling Solution Type Buffer
NIST Traceable Yes
Odor Odorless
Optical Density 1 g/cm3
Packaging Bottle
pH Range 10.01
Quantity 500 mL
Solubility Water Soluble