Learn More
Calcium Lactate Pentahydrate, TRC
Click to view available options
Quantity:
100 g
Unit Size:
100g
Specifications
Specifications
| Chemical Name or Material | Calcium Lactate Pentahydrate |
| CAS | 5743-47-5 |
| Molecular Formula | 2 C3 H5 O3 . Ca . 5 H2 O |
| Quantity | 100 g |
| Synonym | Calcium lactate pentahydrate, Calcium bis(2-hydroxypropanoate) pentahydrate, Propanoic acid, 2-hydroxy-, calcium salt, hydrate (2:1:5), Lactic acid, calcium salt (2:1), pentahydrate (8CI), Propanoic acid, 2-hydroxy-, calcium salt (2:1), pentahydrate (9CI) |
| InChI Formula | InChI=1S/2C3H6O3.Ca.5H2O/c2*1-2(4)3(5)6;;;;;;/h2*2,4H,1H3,(H,5,6);;5*1H2/q;;+2;;;;;/p-2 |
| SMILES | O.O.O.O.O.[Ca+2].CC(O)C(=O)[O-].CC(O)C(=O)[O-] |
| IUPAC Name | calcium;2-hydroxypropanoate;pentahydrate |
| Molecular Weight (g/mol) | 308.294 |
| Formula Weight | 308.063 |
Product Content Correction
Product Title
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur