missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium Lactate Gluconate (Mixture), TRC
Click to view available options
Quantity:
5 g
Unit Size:
5g
Specifications
Specifications
| Chemical Name or Material | Calcium Lactate Gluconate (Mixture) |
| Molecular Formula | 4(C3H5O3) • 2( C6H11O7) • 3( Ca) |
| Quantity | 5 g |
| Synonym | Calcium Gluconolactate, (Gluconato)(lactato)calcium, 2-Hydroxy-propanoic Acid Calcium Complex |
| InChI Formula | InChI=1S/2C6H12O7.4C3H6O3.3Ca/c2*7-1-2(8)3(9)4(10)5(11)6(12)13;4*1-2(4)3(5)6;;;/h2*2-5,7-11H,1H2,(H,12,13);4*2,4H,1H3,(H,5,6);;;/q;;;;;;3*+2/p-6/t2*2-,3-,4+,5-;;;;;;;/m11......./s1 |
| SMILES | [O-]C([C@H](O)C)=O.[O-]C([C@H](O)[C@@H](O)[C@H](O)[C@H](O)CO)=O.[3 Ca2+] |
| IUPAC Name | Di((2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate) tetra ((R)-2-hydroxypropanoate) Tri calsium(II) salt |
| Molecular Weight (g/mol) | 866.82 |
| Formula Weight | 866.06 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction