missing translation for 'onlineSavingsMsg'
Learn More

Calcium chloride, Dihydrate, MP Biomedicals™

Product Code. 11476511 Shop All MP Biomedical Products
Click to view available options
Quantity:
100 g
500 g
1 kg
5 kg
Unit Size:
100g
1kg
500g
5kg

Product Code. 11476511

Brand: MP Biomedicals 0219481590

Please to purchase this item. Need a web account? Register with us today!

Calcium chloride, Dihydrate Cell Culture Reagent, ∽99%, White Powder

  • Calcium chloride is a commonly used reagent in biochemistry.
  • It plays important roles in many biological processes, including signal transduction, muscle contraction, and maintenance of cell membrane and cell wall stability.
  • Calcium chloride is used in the preparation and transformation of competent E. coli and in the transfection of eukaryotic cells with either plasmid DNA or high molecular weight genomic DNA. has been used in the stabilization and two dimensional crystallization of the NADH-ubiquinone oxidoreductase (complex I) from Escherichia coli.
  • It is also used as a dessicant, coagulant in rubber manufacturing and as an accelerator in the concrete industry.
  • Calcium Chloride is a salt of calcium and chloride.
  • It behaves as a typical ionic halide, and is solid at room temperature
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10035-04-8
MDL Number MFCD00149613
Synonym Calcium dichloride dihydrate

Specifications

Melting Point 176°C (Decomposition)(Literature)
pH 4 to 9.5 (5% aq soln)
Physical Form Powder
Quantity 500 g
Assay Percent Range ∽99%
Molecular Formula CaCl2·2H2O
Solubility Information Soluble in water - 294mg/mL (clear, colorless solution) with heat liberation, ethanol, acetic acid, acetone.
Molecular Weight (g/mol) 147
Formula Weight 147
Grade ∽99%
Chemical Name or Material Calcium Chloride Dihydrate
Show More Show Less
compliance-icons
Product Identifier
  • Calcium Chloride Dihydrate, molecular biology reagent
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Calcium Chloride Dihydrate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.