missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melford Calcium Chloride Anhydrous
Click to view available options
Quantity:
1 kg
5 kg
Unit Size:
1kg
5kg
Description
Product usually ships within 24 hours. Research or further manufacturing use only, not for food or drug use.
CAS Number: 10043-52-4
Molecular Formula: CaCl2
Molecular Weight: 110.98
Specifications
Specifications
| Chemical Name or Material | Calcium Chloride Anhydrous |
| CAS | 10043-52-4 |
| Quantity | 1 kg |
| Molecular Formula | CaCl2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction