Learn More
Calcium 3-hydroxy-3-methylbutyrate hydrate, 97+%
Description
Calcium 3-hydroxy-3-methylbutyrate hydrate is a nutritional supplement, which represents a calcium salt of beta-hydroxy beta-methylbutyrate (HMB). The latter is a leucine metabolite produced in the body through oxidation of the ketoacid of leucine (ketoisocaproate, KIC). Because leucine plays an important regulatory role in protein metabolism and due to the conversion of leucine to KIC, several researchers have presumed that KIC is the active ingredient responsible for most of the beneficial effects of leucine on protein metabolism.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | White |
| Quantity | 1 kg |
| Solubility Information | Soluble in water. |
| IUPAC Name | calcium;3-hydroxy-3-methylbutanoic acid |
| Formula Weight | 274.33 (Anhydrous) |
| Percent Purity | ≥97% |
| Physical Form | Powder |
| Chemical Name or Material | Calcium 3-hydroxy-3-methylbutyrate hydrate |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.