Learn More
Caframo™ Stirrer Clamp
For securing Caframo overhead stirrers to A110 (14-500-2) or A210 (14-501-3) stands
Brand:  Caframo™ A120
Product Code. 12907106
  • 151.00€ / Each
Only null left
Estimated Shipment: 20-01-2026
Add to basket
Requires : Overhead stirrers: BDC1850, BDC3030 or BDC6015 (A110 stand) or BDC2002 or BDC2010 (A210 stand).
Description
Description
  • Cast zinc-aluminum alloy coated with a chemical resistant epoxy paint
  • Supports stirrer on the rod of the stand
  • Accepts 0.5 in. diameter stirrer support rod; fits onto stand rods from 0.59 to 1 in. diameter
  • Includes place to hold chuck key
  • Weight: 1.5 lb. (640g)