missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Caffeine 13C3 (trimethyl 13C3) 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Specifications
Specifications
| Chemical Name or Material | Caffeine-13C3 |
| CAS | 78072-66-9 |
| Solvent or Matrix | Acetonitrile |
| Molecular Formula | *C3C5H10N4O2 |
| Quantity | 1 mL |
| Synonym | 3,7-Dihydro-1,3,7-tri(methyl-13C)-1H-purine-2,6-dione, 1H-Purine-2,6-dione, 3,7-dihydro-1,3,7-tri(methyl-13C)-, Caffeine 13C3 (Tirmethyl 13C3) |
| InChI Formula | InChI=1S/C8H10N4O2/c1-10-4-9-6-5(10)7(13)12(3)8(14)11(6)2/h4H,1-3H3/i1+1,2+1,3+1 |
| SMILES | [13CH3]N1C(=O)N([13CH3])c2ncn([13CH3])c2C1=O |
| IUPAC Name | 1,3,7-tri((1^{13}C)methyl)purine-2,6-dione |
| Molecular Weight (g/mol) | 197.17 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction