Learn More
Cadmium Acetate Dihydrate, purum p.a., ≥98.0% (KT), Honeywell™
Product Code. p-7120271 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
10 g
100 g
500 g
Packaging:
Poly Bottle
Unit Size:
100g
10g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15641880 10 g Poly Bottle 10g
15631880 100 g Poly Bottle 100g
15651880 500 g Poly Bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15641880 Supplier Honeywell Research Chemicals Supplier No. 2090110G
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Purum p.a.,≥98.0%
  • Products with a guarantee certificate and/or suitable for the stated analytical application.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 5743-04-4
Molecular Formula C4H10CdO6
Molecular Weight (g/mol) 266.53
MDL Number MFCD00150020
InChI Key AUIZLSZEDUYGDE-UHFFFAOYSA-L
Specifications
Melting Point 256°C
Physical Form Solid
Quantity 10 g
Linear Formula Cd(CH3COO)2 · 2H2O
UN Number UN3077
Beilstein 3730567
Formula Weight 266.53g/mol
Percent Purity ≥98.0% (KT)
Grade purum p.a.
Packaging Poly Bottle
Density 2.01g/cu.cm
Chemical Name or Material Cadmium Acetate Dihydrate