Learn More
Cadence Science™ Laboratory Pipetting Needles
Product Code. 11774726
Click to view available options
Needle Gauge:
14 G
16 G
18 G
20 G
22 G
24 G
Length (English) Needle:
1 in.
1.5 in.
2 in.
4 in.
6 in.
Material:
Nickel-plated (Hub), Stainless Steel
Stainless Steel, Stainless Steel (Hub)
Connection Type:
Female Luer
Luer
Unit Size:
Pack of 12
Product Code. 11774726
Brand: Cadence Science™ 7932
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
316 Stainless Steel Hub with 316 Stainless Steel Tubing
  • Needles 14-825-16A through -16N and 14-818-106A through -106C all have standard nickel plated hubs and 304 stainless steel cannulae
  • Needles 01-290-29 through -35 have 316 stainless steel Micro-Mate™ Threaded Female Luer Hubs and 316 stainless steel cannulae
  • All have 90° blunt ends
Specifications
Material Nickel-plated (Hub), Stainless Steel
Needle Gauge 20 G
Length (English) Needle 1.5 in.
Product Type Pipetting Needle
Quantity 12 Needles
Needle Point Style 90° Blunt
For Use With (Equipment) Manual and Automatic Volumetric Dispensers, Diluters, Pipetters
Connection Type Luer