Learn More
Cadence Science™ Deflected Noncoring Septum Penetration Needles
Click to view available options
Needle Gauge:
18 G
19 G
20 G
21 G
22 G
25 G
Length (English) Needle:
1 in.
1.5 in.
2 in.
4 in.
6 in.
8 in.
12 in.
Unit Size:
Pack of 12
Description
Use in gas chromatography and nuclear magnetic resonance spectroscopy procedures
- Penetration needles with deflected noncoring septum
- For noncoring penetration of septa and rubber closures involving air-sensitive and pyrophoric compounds
Specifications
Specifications
| Material | Nickel-plated Brass (Hub), Stainless Steel (Tube) |
| Needle Gauge | 18 G |
| For Use With (Application) | Use in gas chromatography and nuclear magnetic resonance spectroscopy procedures |
| Includes | Deflected non-coring septum |
| Length (English) Needle | 6 in. |
| Product Type | Penetration Needle |
| Quantity | 12 Needles |
| Needle Point Style | Deflected End |