Learn More
Cadence Science™ Deflected Noncoring Septum Penetration Needles
Product Code. 11755905
Click to view available options
Needle Gauge:
18 G
19 G
20 G
21 G
22 G
25 G
Length (English) Needle:
1 in.
1.5 in.
2 in.
4 in.
6 in.
8 in.
12 in.
Unit Size:
Pack of 12
Product Code. 11755905
Brand: Cadence Science™ 4193
Use in gas chromatography and nuclear magnetic resonance spectroscopy procedures.
Use in gas chromatography and nuclear magnetic resonance spectroscopy procedures
  • Penetration needles with deflected noncoring septum
  • For noncoring penetration of septa and rubber closures involving air-sensitive and pyrophoric compounds
Specifications
Material Nickel-plated Brass (Hub), Stainless Steel (Tube)
Needle Gauge 18 G
For Use With (Application) Use in gas chromatography and nuclear magnetic resonance spectroscopy procedures
Includes Deflected non-coring septum
Length (English) Needle 2 in.
Product Type Penetration Needle
Quantity 12 Needles
Needle Point Style Deflected End