Description
- Suitable for use with a C6 Plus Flow Cytometer
- Includes the following:
- Peristalitc pump tubing (12)
- Sheath bottle filters (6)
- In-line sheath filters (6)
Specifications
Specifications
| Includes | Peristaltic pump tubing (12), sheath bottle filters (6), Ini-line sheath filters (6) |
| Quantity | Each |
| For Use With (Equipment) | C6 Plus Flow Cytometer |
| Product Type | Maintenance Kit |