Learn More
Uvex™ C500 Cut-Resistant Sleeve
Product Code. p-7236927 Shop All Uvex Products
Change view
Click to view available options
Size:
Medium
Large
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
15327042 Medium Each
15317042 Large Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15327042 Supplier Uvex™ Supplier No. 6049107
Bamboo-TwinFlex™ Technology ensures a high level of cut protection and abrasion resistance. Silky, soft bamboo fiber increases comfort and moisture absorption.
  • Bamboo-TwinFlex™ Technology ensures cut protection and abrasion resistance
  • Bamboo fiber is soft and silky for increased comfort
  • Knit cuff offers a secure fit
  • Silicone-free according to the imprint test
  • Ideal for applications in the automotive, construction, beverage, glass, and metal industries
TRUSTED_SUSTAINABILITY
Specifications
Size Medium
Product Type Cut-resistant Sleeve
Material Bamboo Viscose, HPPE, Polyamide
Color Green
Sterility Non-sterile
Style Standard Sleeve
Disposable Reusable
Closure Type Elastic Ends
Seam Type Taped
Cuff Style Knit