Learn More
Novus Biologicals™ C32 Nuclear Cell Lysate
Product Code. 18516181 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18516181 0.05 mg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18516181 Supplier Novus Biologicals™ Supplier No. L002V4
 Restricted Item
Estimated Shipment: 14-10-2026
Nuclear Extract Lysate
TRUSTED_SUSTAINABILITY
Specifications
Preparation Method Use it directly for immuno-precipitation, or heat lysate with SDS gel loading buffer to 95C for 5 minutes followed by rapid cooling for western blot application. If dissociating conditions are required, add reducing agent prior to heating.
Quantity 0.05 mg
Content And Storage Store at -80C. Avoid freeze-thaw cycles.
Target C32