Learn More
Consort C3010/C3030 Multiparameter Dual Channel Analyzers
Product Code. 11792009
Click to view available options
Product Type:
Multiparameter Analyzer
Unit Size:
Each
Product Code. 11792009
Brand: Consort C3030
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Two independent channels for all measurements. Consort C3010/C3030 Multiparameter Dual Channel Analyzers are full parameter benchtop meters with unique measurement hardware for each channel, separate temperature inputs, data logging and a USB/RS232 interface for connection to/control by a PC.
  • Multichannel up to 2 measurements can be performed at the same time (2X/sec)
  • Multiparameter for pH, mV, conductivity, resistivity, salinity, TDS, dissolved oxygen, temperature, ion (3030 only), and air pressure
  • No interference between pH/ORP/Ion and conductivity electrodes in the same solution
  • Custom calibration tables for buffer/standard-temperature relations (via device menu or from PC)
  • Wide display shows 2 channels at the same time, including temperature and date/time
  • Stability indicator ensures visualization when the measurement has stabilized
  • Stability algorithm ensures stable readings with ability to detect fast changes
  • Gold plated BNC connectors each accept different electrodes; gold plating prevents corrosion
  • Hold function allows to freeze the display
  • Selectable resolution for more stable readings of mV, pH and DO
  • Range lock for conductivity measurements
  • Built-in help system helps guide users through all steps of calibration and use
  • Capacitive compensation eliminates electrode/cable capacitive component at low conductivities
  • Galvanic isolated USB interface eliminates ground loop effects when connected to a PC
  • GPL report can be shown on the display or sent to the digital port
  • Pre-programmed standards:
    pH: 1.68, 2.00, 4.00, 4.01, 6.87, 6.99, 9.18, 9.21, 10.01, 12.00, 12.45 (at 25°C)
    Conductivity: 1413 μS/cm, 12.88 mS/cm, 111.8 mS/cm (at 25°C)
  • Free software and firmware updates downloadable from www.consort.be
Specifications
Outputs USB/RS232
Accuracy (Conductivity) 0.5% f.s. of range
Accuracy (DO) 1% ± 1 digit
Accuracy (ISE) 0.5% ± 1 digit
Accuracy (mV) 0.1% ± 1 digit
Accuracy (pH) 0.1% ±1 digit
Backlight Option White back light
Calibration GLP
Calibration Points 1 (mV, DO, temperature, air pressure)
Calibration Points (Conductivity) 1 to 3
Calibration Points (ISE) 2 to 5 points + blank
Calibration Points (pH) 1 to 5
Cell Constant 0.07 to 13 cm-1
Data Management Details Measurement + ° C/date/time
Data Points 12,000
Channels 2 measurement, 2 temperature
Display Type LCD, 240 x 64 pixel
Range (Conductivity) 0 to 2000 mS/cm
Range (mV) ±2000.0 mV
Range (pH) -2.000 to 16.000
Range (Resistivity) 0 to 200 Mohm/cm
Range (Salinity) 0.0 to 70.0
Range (TDS) 0.0 to 100.0 g/L
Resolution (ISE) 3 digits
Resolution (pH) 0.001
Temperature Range (Metric) -5.0°C to +105.0°C
For Use With (Application) ion, pH, mV, conductivity, resistivity, salinity, TDS, DO, air pressure, temperature
Includes C3030 Meter Only
Inputs 2 BNC (measurement), 2 x 2 banana (for Pt1000 for temperature)
Languages English, Dutch, French, German
Model C3030
Pressure Range C3030
Product Type Multiparameter Analyzer
Range (DO) 0.00 - 60.00 mg/L
Range (ISE) 0.01ng/L to 100 g/L
Relative Humidity Range 0 - 95%, non-condensing
Resolution (Conductivity) 0.001 μS/cm
Resolution (DO) 0.01 mg/L
Resolution (mV) 0.1 mV
Resolution (Resistivity) 1 Ω.cm
Resolution (TDS) 0.01 mg/L
Salinity Factor Ref temp: 15° C
Temperature Accuracy 0.1°C
Temperature Compensation −5.0° to +105.0° C (pH, conductivity), 0° to 50° C (DO)
Temperature (Metric) Operating 0°C to 40°C
Temperature (Metric) Reference 20°/25° C (conductivity)
Temperature Resolution 0.1° C
Warranty 36 Months
Dimensions (L x W x H) 26W x 18D x 9H cm
Weight (Metric) 1 kg
Voltage 100/240 V
Frequency 50/60 Hz