missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ C20orf81 Recombinant Protein
Brand: Abnova™ H00064773-P01.25ug
Additional Details : Weight : 0.00010kg
Description
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Specifications
AAH37240 | |
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer | |
75.68 | |
8 | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
MVFCLSSEEPRRPLRSDMVHFQASEVQQLLHNKFVVILGDSIQRAVYKDLVLLLQKDSLLTAAQLKAKGELSFEQDQLVAGGQLGELHNGTQYREVRQFCSGSGHHLVRFYFLTRVYSEYLEDVLEELTYGPAPDLVIINSCLWDLSRYGRCSMESYRENLERVFVRMDQVLPDSCLLVWNMAMPLGERITGGFLLPELQPLAGSLRRDVVEGNFYSATLAGDHCFDVLDLHFHFRHAVQHRHRDGVHWDQHAHRHLSHLLLTHVADAWGVELPKRGYPPDPWIEDWAEMNHPFQGSHRQTPDFGEHLALLPPPPSSLPPPMPFPYPLPQPSPPPLFPPLPQDTPFFPGQPFPPHEFFNYNPVEDFSMPPHLGCGPGVNFVPGPLPPPIPGPNPHGQHWGPVVHRGMPRYVPNSPYHVRRMGGPCRQRLRHSERLIHTYKLDRRPPAHSGTWPG | |
C20orf81/DKFZp547L054/FLJ22376/bA12M19.1 | |
FAM113A | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Protein Array, ELISA, Western Blot | |
64773 | |
C20orf81 (Human) Recombinant Protein (P01) | |
In vitro wheat germ expression system | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Wheat Germ (in vitro) | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FAM113A | |
Human | |
Recombinant | |
Solution |