missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C19orf54 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA544437
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: MTSPCSPPLK PPISPPKTPV PQASSIPSPP LPPSPLDFSA LPSPPWSQQT Sequence homology: Human: 100%
C19orf54 Polyclonal antibody specifically detects C19orf54 in Human samples. It is validated for Western Blot.Specifications
C19orf54 | |
Polyclonal | |
Unconjugated | |
C19orf54 | |
C19orf54; chromosome 19 open reading frame 54; UPF0692 protein C19orf54 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
284325 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q5BKX5 | |
C19orf54 | |
Synthetic peptide directed towards the N-terminal of human C19orf54 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |