Learn More
C16orf72 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA559626
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: KLWHLFQNSA TAVAQLYKDR VCQQPGLSLW VPFQNAATAV TNLYKESVDT HQRSFDIGIQ IGYQRRNKDV LAWVKKRRRT IRREDLISFL C Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
C16orf72 is a protein coding gene.Specifications
C16orf72 | |
Polyclonal | |
Unconjugated | |
C16orf72 | |
C16orf72; chromosome 16 open reading frame 72; PRO0149; UPF0472 protein C16orf72 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
29035 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q14CZ0 | |
C16orf72 | |
Recombinant protein corresponding to Human C16orf72 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only