Learn More
C12E8, TRC
Specifications
Specifications
| Chemical Name or Material | 2-[2-[2-[2-[2-[2-[2-(2-dodecoxyethoxy)ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethanol |
| CAS | 3055-98-9 |
| Molecular Formula | C28H58O9 |
| Quantity | 250 mg |
| Synonym | C12E8 |
| InChI Formula | InChI=1S/C28H58O9/c1-2-3-4-5-6-7-8-9-10-11-13-30-15-17-32-19-21-34-23-25-36-27-28-37-26-24-35-22-20-33-18-16-31-14-12-29/h29H,2-28H2,1H3 |
| SMILES | CCCCCCCCCCCCOCCOCCOCCOCCOCCOCCOCCOCCO |
| IUPAC Name | 2-[2-[2-[2-[2-[2-[2-(2-dodecoxyethoxy)ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethanol |
| Molecular Weight (g/mol) | 538.75 |
| Formula Weight | 538.4081 |