Learn More
Novus Biologicals™ C-Reactive Protein/CRP Antibody Pair [HRP]
Shop All Bio Techne Products
Click to view available options
Quantity:
15 Plates
5 Plates
Unit Size:
Pack of 15
Pack of 5
Description
This solid phase sandwich ELISA utilizes an antibody specific for Mouse C-Reactive Protein/CRP.
Specifications
Specifications
| Assay Range | 34.38 pg/mL |
| Assay Sensitivity | 34.38-2200 pg/mL |
| Assay | Sandwich ELISA |
| Conjugate | HRP |
| Immunoassay Kit Format | Sandwich ELISA Kit |
| Product Type | Antibody Pairs |
| Specificity | This solid phase sandwich ELISA utilizes an antibody specific for Mouse C-Reactive Protein/CRP. |
| Target Species | Mouse |
| For Use With (Application) | Sandwich ELISA |
| Gene Alias | C-reactive protein, C-reactive protein, pentraxin-related, MGC88244, pentraxin 1, PTX1MGC149895 |
| Show More |