Learn More
Wheaton™ C Model Roller Culture Apparatus
Product Code. 15254287 Shop All Wheaton Science Products Products
Change view
Click to view available options
No. of Positions:
55
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Positions Unit Size
15254287 55 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15254287 Supplier Wheaton™ Supplier No. WRBPR5110C
  • ONLINE EXCLUSIVE PRICE
    25395.00€
    19042.98€ / Pack of 1
    Save 6352.02€
    25% Off
 Restricted Item
Estimated Shipment: 04-01-2027
Specifications
Speed Range 0.25 to 8.1 rpm
Electrical Requirements 100/240 V, 50/60 Hz, 35 W
Voltage 100/240 V
Temperature Range (Metric) 10°C to 40°C
Wattage 35 W
Certifications/Compliance CE, CSA, UL
Frequency 50/60 Hz
Model C Model
For Use With (Application) For Adherent Cell Culture
Humidity Range 80% up to 31°C
No. of Positions 55
Packaging Quantity 1 Ea.
Spacing 15.2 in.