Learn More
Wheaton™ C Model Roller Culture Apparatus
Product Code. 13433866 Shop All Wheaton Science Products Products
Click to view available options
No. of Positions:
52
Unit Size:
Each
Product Code. 13433866
Brand: Wheaton™ WRTPF5110C
Only null left
Add to basket
Add to basket
For use in laboratory, pilot plant and production laboratory cell culture installations. Wheaton™ C Model Roller Culture Apparatus provides several configuration options.
  • Choice of configuration
TRUSTED_SUSTAINABILITY
Specifications
Speed Range 0.25 to 8.1 rpm
Electrical Requirements 100/240 V, 50/60 Hz, 35 W
Voltage 100/240 V
Temperature Range (Metric) 10°C to 40°C
Wattage 35 W
Certifications/Compliance CE, CSA, UL
Frequency 50/60 Hz
Depth (Metric) 63 cm
Height (Metric) 190 cm
Model C Model
For Use With (Application) For Adherent Cell Culture
Humidity Range 80% up to 31°C
No. of Positions 52
Packaging Quantity 1 Ea.