Learn More
Wheaton™ C Model R2P 2.0 Standard Roller Culture Apparatus with Temperature Monitoring and Battery Backup
Product Code. 13463886 Shop All Wheaton Science Products Products
Change view
Click to view available options
No. of Positions:
15
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Positions Unit Size
13463886 15 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 13463886 Supplier Wheaton™ Supplier No. WRBMR5031C
  • ONLINE EXCLUSIVE PRICE
    15277.50€
    11457.35€ / Pack of 1
    Save 3820.15€
    25% Off
 Restricted Item
Estimated Shipment: 19-01-2027
Specifications
Electrical Requirements 230 V, 50/60 Hz, 35 W
Certifications/Compliance CE, CSA, ROHS, UL
Frequency 50/60 Hz
Model C-Model
For Use With (Application) For system navigation For Types of Screens
Humidity Range 80% up to 31°C
No. of Positions 15