Learn More
Wheaton™ C Model R2P 2.0 Standard Roller Culture Apparatus
Brand:  Wheaton™ WRBMR5010-C
8502.50 EUR valid until 2024-12-31
Use promo code "23261" to get your promotional price.
Additional Details : Weight : 40.82330kg
Product Code. 13433886
  • 8950.00€ / Each
Estimated Shipment: 05-08-2024
Specifications
0.25 to 8.1 rpm
230 V
35 W
50/60 Hz
34 cm
35 kg
For system navigation For Types of Screens
1 Ea.
230 V, 50/60 Hz, 35 W
40°C
CE, CSA, ROHS, UL
63 cm
76 cm
C-Model
80% up to 31°C