Learn More
Wheaton™ C Model R2P 2.0 Roller Culture Apparatus With Temperature Monitoring and Battery Backup
Product Code. 13413886 Shop All Wheaton Science Products Products
Change view
Click to view available options
No. of Positions:
55
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Positions unitSize
13413886 55 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 13413886 Supplier Wheaton™ Supplier No. WRBPF5111C
Specifications
Speed Range 0.25 to 8.1 rpm
Electrical Requirements 100/240 V, 50/60 Hz, 35 W
Voltage 100/240 V
Temperature Range (Metric) 10°C to 40°C
Wattage 35 W
Certifications/Compliance CE, CSA, UL
Frequency 50/60 Hz
Model C Model
For Use With (Application) For Adherent Cell Culture
Humidity Range 80% up to 31°C
No. of Positions 55
Packaging Quantity 1 Ea.
Spacing 15.2 in.