Learn More
Wheaton™ C Model Bottom Drive Modular Spacing Roller Culture Apparatus with Temperature Monitoring and Battery Backup
Product Code. 13443896 Shop All Wheaton Science Products Products
Change view
Click to view available options
No. of Positions:
45
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Positions Unit Size
13443896 45 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 13443896 Supplier Wheaton™ Supplier No. WRBMR5091C
  • ONLINE EXCLUSIVE PRICE
    26444.25€
    25166.21€ / Pack of 1
    Save 1278.04€
    4% Off
 Restricted Item
Estimated Shipment: 30-11-2026
Specifications
Speed Range 0.25 to 8.1 rpm
Electrical Requirements 100/240 V, 50/60 Hz, 35 W
Voltage 100/240 V
Temperature Range (Metric) 10°C to 40°C
Wattage 35 W
Certifications/Compliance CE, CSA, UL
Frequency 50/60 Hz
Model C Model
For Use With (Application) For laboratory, pilot plant and production cell culture installations
Humidity Range 80% up to 31°C
No. of Positions 45
Packaging Quantity 1 Ea.
Spacing 17.7 cm
Show More Show Less
Product Title
Sélectionnez un problème
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.