missing translation for 'onlineSavingsMsg'
Learn More

[C(E)]-N-[(6-Chloro-3-pyridinyl)methyl]-N'-nitro-guanidine, TRC

Product Code. 30355697
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30355697 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30355697 Supplier Toronto Research Chemicals Supplier No. TRCC38089050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material [C(E)]-N-[(6-Chloro-3-pyridinyl)methyl]-N'-nitro-guanidine
CAS 1575569-69-5
Molecular Formula C7H8ClN5O2
Quantity 50 mg
Synonym [C(E)]-N-[(6-Chloro-3-Pyridinyl)Methyl]-N’-Nitro-Guanidine
InChI Formula InChI=1S/C7H8ClN5O2/c8-6-2-1-5(3-10-6)4-11-7(9)12-13(14)15/h1-3H,4H2,(H3,9,11,12)
SMILES N\C(=N/[N+](=O)[O-])\NCc1ccc(Cl)nc1
IUPAC Name 1-[(6-chloropyridin-3-yl)methyl]-2-nitroguanidine
Molecular Weight (g/mol) 229.62
Formula Weight 229.0367
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.