missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BzATP Ammonium Salt (Mixture of 2'-3' isomers), TRC
Specifications
Specifications
| Chemical Name or Material | BzATP Ammonium Salt (Mixture of 2'-3' isomers) |
| Molecular Formula | C24H24N5O15P3 xNH3 |
| Quantity | 5 mg |
| Synonym | 2'(3')-O-(4-Benzoylbenzoyl)adenosine 5′-triphosphate Ammonium Salt |
| InChI Formula | InChI=1S/2C24H24N5O15P3/c25-21-17-22(27-11-26-21)29(12-28-17)23-19(31)20(16(41-23)10-40-46(36,37)44-47(38,39)43-45(33,34)35)42-24(32)15-8-6-14(7-9-15)18(30)13-4-2-1-3-5-13;25-21-17-22(27-11-26-21)29(12-28-17)23-20(19(31)16(41-23)10-40-46(36,37)44-47(38,39)43-45(33,34)35)42-24(32)15-8-6-14(7-9-15)18(30)13-4-2-1-3-5-13/h2*1-9,11-12,16,19-20,23,31H,10H2,(H,36,37)(H,38,39)(H2,25,26,27)(H2,33,34,35)/t2*16-,19-,20-,23-/m11/s1 |
| SMILES | NC1=NC=NC2=C1N=CN2[C@H](O3)[C@H](OC(C4=CC=C(C(C5=CC=CC=C5)=O)C=C4)=O)[C@H](O)[C@H]3CO[P](O[P](O[P](O)(O)=O)(O)=O)(O)=O.O=C(O[C@H]6[C@@H](O)[C@H](N7C(N=CN=C8N)=C8N=C7)O[C@@H]6CO[P](O[P](O[P](O)(O)=O)(O)=O)(O)=O)C9=CC=C(C(C%10=CC=CC=C%10)=O)C=C9 |
| IUPAC Name | [(2R,3R,4R,5R)-2-(6-aminopurin-9-yl)-4-hydroxy-5-[[hydroxy-[hydroxy(phosphonooxy)phosphoryl]oxyphosphoryl]oxymethyl]oxolan-3-yl] 4-benzoylbenzoate;[(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-4-hydroxy-2-[[hydroxy-[hydroxy(phosphonooxy)phosphoryl]oxyphosphoryl]oxymethyl]oxolan-3-yl] 4-benzoylbenzoate |
| Molecular Weight (g/mol) | 715.39 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?