Learn More
BW B70C, TRC
Specifications
Specifications
| Chemical Name or Material | 1-[(E)-4-[3-(4-fluorophenoxy)phenyl]but-3-en-2-yl]-1-hydroxyurea |
| CAS | 134470-38-5 |
| Molecular Formula | C17 H17 F N2 O3 |
| Quantity | 25 mg |
| Synonym | BW B70C, N-[3-[3-(4-Fluorophenoxy)phenyl]-1-methyl-2-propen-1-yl]-N-hydroxyurea, BW-B 70C, N-[3-[3-(4-fluorophenoxy)phenyl]-1-methyl-2-propenyl]-N-hydroxy-urea |
| InChI Formula | InChI=1S/C17H17FN2O3/c1-12(20(22)17(19)21)5-6-13-3-2-4-16(11-13)23-15-9-7-14(18)8-10-15/h2-12,22H,1H3,(H2,19,21)/b6-5+ |
| SMILES | CC(\C=C\c1cccc(Oc2ccc(F)cc2)c1)N(O)C(=O)N |
| IUPAC Name | 1-[(E)-4-[3-(4-fluorophenoxy)phenyl]but-3-en-2-yl]-1-hydroxyurea |
| Molecular Weight (g/mol) | 316.327 |
| Formula Weight | 316.122 |