Learn More
Butyryl-L-carnitine-d3 HCl (N-methyl-d3), CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | L-Carnitine: HCl, o-butyryl (N-methyl-D3) |
| CAS | 1334532-21-6 |
| Molecular Formula | C11 D3 H19 N O4 . Cl |
| Quantity | 0.01 g |
| Synonym | 1-Propanaminium, 3-carboxy-N,N-dimethyl-N-(methyl-d3)-2-(1-oxobutoxy)-, chloride (1:1), (2 R)- (ACI), (3 R)-3-Butanoyloxy-4-(dimethyl-(trideuteriomethyl)azaniumyl)butanoate hydrochloride, Butyryl-L-carnitine-d3 Chloride, Butyryl-L-carnitine-D3 (N-methyl-D3) chloride, (2 R)-3-Carboxy-N,N-dimethyl-N-(methyl-d3)-2-(1-oxobutoxy)-1-propanaminium chloride, [(2 R)-3-Carboxy-2-(butanoyloxy)propyl]dimethyl(methyl-d3)ammonium chloride |
| InChI Formula | InChI=1 S/C11H21NO4.ClH/c1-5-6-11(15)16-9(7-10(13)14)8-12(2,3)4;/h9H,5-8H2,1-4H3;1 H/t9-;/m1./s1/i2D3; |
| SMILES | [Cl-].[2 H]C([2 H])([2 H])[N+](C)(C)C[C@@H](CC(=O)O)OC(=O)CCC |
| IUPAC Name | [(2 R)-2-butanoyloxy-3-carboxypropyl]-dimethyl-(trideuteriomethyl)azanium;chloride |
| Molecular Weight (g/mol) | 270.7682 |
| Formula Weight | 270.1426 g/mol |
| Show More |