missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Butyryl-d7-L-carnitine HCl, CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | Butyryl-L-carnitine-D7 (butyryl-D7) Chloride |
| Molecular Formula | C11 D7 H15 N O4 . Cl |
| Quantity | 0.05 g |
| Synonym | Butyryl-d7-L-carnitine HCl, Butyryl-L-carnitine-d7 chloride, Butyryl-L-carnitine-D7 (butyryl-D7) chloride, (2 R)-3-Carboxy-N,N,N-trimethyl-2-(1-oxobutoxy-2,2,3,3,4,4,4-d7)-1-propanaminium chloride, [(2 R)-3-Carboxy-2-(butanoyloxy-d7)propyl]trimethylammonium chloride |
| InChI Formula | InChI=1 S/C11H21NO4.ClH/c1-5-6-11(15)16-9(7-10(13)14)8-12(2,3)4;/h9H,5-8H2,1-4H3;1 H/t9-;/m1./s1/i1D3,5D2,6D2; |
| SMILES | [Cl-].[2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C(=O)O[C@H](CC(=O)O)C[N+](C)(C)C |
| IUPAC Name | [(2 R)-3-carboxy-2-(2,2,3,3,4,4,4-heptadeuteriobutanoyloxy)propyl]-trimethylazanium;chloride |
| Molecular Weight (g/mol) | 274.7928 |
| Formula Weight | 274.1677 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction