missing translation for 'onlineSavingsMsg'
Learn More

Butyric-4,4,4-d3 Acid, TRC

Product Code. 30335779
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30335779 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30335779 Supplier Toronto Research Chemicals Supplier No. TRCB69224110MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Butanoic Acid 4,4,4-D3
CAS 36789-14-7
Molecular Formula C4H5D3O2
Quantity 10 mg
Synonym Butanoic-4,4,4-d3 acid (9CI), Butyric-4,4,4-d3 acid, Butanoic acid 4,4,4-D3
InChI Formula InChI=1S/C4H8O2/c1-2-3-4(5)6/h2-3H2,1H3,(H,5,6)/i1D3
SMILES OC(CCC([2H])([2H])[2H])=O
IUPAC Name butanoic-4,4,4-d3 acid
Molecular Weight (g/mol) 91.12
Formula Weight 91.07
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.