Learn More
Wheaton™ Rubber Freeze Dry Stoppers
Product Code. 14833592 Shop All Wheaton Science Products Products
Click to view available options
Diameter (Metric):
14 mm
20 mm
Unit Size:
Pack of 1000
Product Code. 14833592
Brand: Wheaton™ 700005
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Provides for safe and secure closure of bottles and vials. Wheaton™ Rubber Freeze Dry Stoppers are manufactured from butyl rubber.
  • Available in two sizes
  • Freeze dry stoppers are push-in (not screw thread)
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric) 20 mm
For Use With (Equipment) Diagnostic vials
Color Gray
Material Butyl Rubber
Product Type Plug Stopper