Learn More
Macherey-Nagel™ Butyl Phase Polygosil™ C4
Product Code. 10661482
Click to view available options
Quantity:
10 g
100 g
Particle Size:
7 μm
Pore Size:
1000 Å
300 Å
Unit Size:
100g
10g
Product Code. 10661482
Brand: Macherey-Nagel™ 711991.10