Learn More
Macherey-Nagel™ Butyl Phase Polygosil™ C4
Click to view available options
Quantity:
10 g
100 g
Particle Size:
7 μm
Pore Size:
1000 Å
300 Å
Unit Size:
100g
10g
Specifications
Specifications
| Particle Size | 7 μm |
| Pore Size | 1000 Å |
| Technique | Liquid Chromatography |
| Quantity | 100 g |
| Type | Polygosil C4 Phases |