missing translation for 'onlineSavingsMsg'
Learn More

Butoxycaine, TRC

Product Code. 30331823
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30331823 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30331823 Supplier Toronto Research Chemicals Supplier No. TRCB80885050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Butoxycaine
CAS 3772-43-8
Molecular Formula C17 H27 N O3
Quantity 50 mg
Synonym Benzoic acid, 4-butoxy-, 2-(diethylamino)ethyl ester (9CI, ACI), Benzoic acid, p-butoxy-, 2-(diethylamino)ethyl ester (6CI, 7CI, 8CI), 2-(Diethylamino)ethyl 4-butoxybenzoate, 2-(Diethylamino)ethyl 4-n-butoxybenzoate, Butoxycaine
InChI Formula InChI=1S/C17H27NO3/c1-4-7-13-20-16-10-8-15(9-11-16)17(19)21-14-12-18(5-2)6-3/h8-11H,4-7,12-14H2,1-3H3
SMILES CCCCOc1ccc(cc1)C(=O)OCCN(CC)CC
IUPAC Name 2-(diethylamino)ethyl 4-butoxybenzoate
Molecular Weight (g/mol) 293.401
Formula Weight 293.199
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.