missing translation for 'onlineSavingsMsg'
Learn More

Butoconazole Nitrate, TRC

Product Code. 30335837
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30335837 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30335837 Supplier Toronto Research Chemicals Supplier No. TRCB6902731G
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Butoconazole Nitrate
CAS 64872-77-1
Molecular Formula C19 H17 Cl3 N2 S . H N O3
Quantity 1 g
Synonym (+/-)-1-[4-(4-Chlorophenyl)-2-[(2,6-dichlorophenyl)thio]butyl]-1H-imidazole mononitrate, 1-[4-(4-Chlorophenyl)-2-[(2,6-dichlorophenyl)thio]butyl]-1H-imidazole mononitrate, (+/-)-Butoconazole nitrate, Butaconazole nitrate, Butoconazole nitrate, Femstat, Gynomyk
InChI Formula InChI=1S/C19H17Cl3N2S.HNO3/c20-15-7-4-14(5-8-15)6-9-16(12-24-11-10-23-13-24)25-19-17(21)2-1-3-18(19)22;2-1(3)4/h1-5,7-8,10-11,13,16H,6,9,12H2;(H,2,3,4)
SMILES O[N+](=O)[O-].Clc1ccc(CCC(Cn2ccnc2)Sc3c(Cl)cccc3Cl)cc1
IUPAC Name 1-[4-(4-chlorophenyl)-2-(2,6-dichlorophenyl)sulfanylbutyl]imidazole;nitric acid
Molecular Weight (g/mol) 474.79
Formula Weight 473.0134
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.