Learn More
Esculape Laboratoire™ Burns Aid Kit
Product Code. 11788383
Change view
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11788383 1 Ea. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11788383 Supplier Esculape Laboratoire™ Supplier No. 3170703
  • ONLINE EXCLUSIVE PRICE
    92.40€
    50.92€ / Pack of 1
    Save 41.48€
    44% Off
Estimated Shipment: 12-11-2026
Specifications
Depth (Metric) 5 cm
Length (Metric) 17 cm
Product Type Burns Aid Kit
Includes Mainly to treat all types of thermal burns, 4 fixing clips for bands, 2 bactericidal soap pockets, 2 antiseptic compresses, 2 anti-blows compresses, 1 roll sticking plaster 5 mx 2cm, 10 adhesive bandages, 1 gauze bandage 3 x 7cm, 1 compression bandage 6cm
Quantity 1 Ea.
Height (Metric) 23 cm