Learn More
Afora™ Burettes with Reservoir
Click to view available options
Capacity (Metric):
10 mL
Unit Size:
Each
Specifications
Specifications
| Capacity (Metric) | 10 mL |
| Material | Glass |
| Graduations | 0.20 mL |
| Tolerance | 0.02 mL |
| Type | Buret |
| Class | Class A |