Learn More
Bupivacaine Hydrochloride Monohydrate, TRC
Specifications
Specifications
| Chemical Name or Material | Bupivacaine Hydrochloride Monohydrate |
| CAS | 73360-54-0 |
| Molecular Formula | C18 H28 N2 O . Cl H . H2 O |
| Quantity | 500 mg |
| Synonym | 2-Piperidinecarboxamide, 1-butyl-N-(2,6-dimethylphenyl)-, hydrochloride, hydrate (1:1:1), 2-Piperidinecarboxamide, 1-butyl-N-(2,6-dimethylphenyl)-, monohydrochloride, monohydrate (9CI), Bupivacaine hydrochloride monohydrate, 1-Butyl-N-(2,6-dimethylphenyl)-2-piperidinecarboxamide monohydrochloride monohydrate |
| InChI Formula | InChI=1S/C18H28N2O.ClH.H2O/c1-4-5-12-20-13-7-6-11-16(20)18(21)19-17-14(2)9-8-10-15(17)3;;/h8-10,16H,4-7,11-13H2,1-3H3,(H,19,21);1H;1H2 |
| SMILES | O.Cl.CCCCN1CCCCC1C(=O)Nc2c(C)cccc2C |
| IUPAC Name | 1-butyl-N-(2,6-dimethylphenyl)piperidine-2-carboxamide;hydrate;hydrochloride |
| Molecular Weight (g/mol) | 342.9 |
| Formula Weight | 342.2074 |