Learn More
Wld Tec™ gasprofi 1 SCS micro Gas Burner
Product Code. 10241202
Change view
Click to view available options
Gas Type:
Butane, Natural Gas, Propane
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Gas Type Unit Size
10241202 Butane, Natural Gas, Propane Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10241202 Supplier Wld Tec™ Supplier No. 6.004.000
  • ONLINE EXCLUSIVE PRICE
    772.80€
    602.00€ / Pack of 1
    Save 170.80€
    22% Off
Estimated Shipment: 19-11-2026
The smallest professional laboratory gas burner. Wld Tec™ Gasprofi 1 SCS Micro Gas Burner offers optimal safety and easy operation.
  • Button or foot pedal operation
  • Gas inlet filter
  • Safety control system (SCS)
  • Removable and decomposable burner head
  • Housing, burner head, gas connection and controls made of stainless steel
  • DVGW approved
Specifications
Gas Type Butane, Natural Gas, Propane
Length (Metric) 85 mm
Material Stainless Steel
Height (Metric) 49 mm
Includes Foot Pedal and Button Function
Max. Temperature (Metric) 1300°C (Natural Gas), 1350°C (Liquid Gas)
No. per Pack 1 Ea.
Weight (Metric) 700 g
Width (Metric) 86 mm
Product Type Bunsen Burner