missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bumadizon, TRC
Specifications
Specifications
| Chemical Name or Material | (2RS)-2-[(1,2-Diphenyldiazanyl)carbonyl]hexanoic Acid |
| CAS | 3583-64-0 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C19 H22 N2 O3 |
| Quantity | 100 mg |
| Synonym | (2RS)-2-[(1,2-Diphenyldiazanyl)carbonyl]hexanoic Acid, Phenylbutazone Imp. A (EP), Propanedioic acid, 2-butyl-, 1-(1,2-diphenylhydrazide), Malonic acid, butyl-, 1,2-diphenylhydrazide (7CI), Malonic acid, butyl-, mono(1,2-diphenylhydrazide) (8CI), Propanedioic acid, butyl-, mono(1,2-diphenylhydrazide) (9CI), Bumadizon, Bumadizone, Butylmalonic acid mono(1,2-diphenylhydrazide), N-(2-Carboxycaproyl)hydrazobenzene |
| InChI Formula | InChI=1S/C19H22N2O3/c1-2-3-14-17(19(23)24)18(22)21(16-12-8-5-9-13-16)20-15-10-6-4-7-11-15/h4-13,17,20H,2-3,14H2,1H3,(H,23,24) |
| SMILES | CCCCC(C(=O)O)C(=O)N(Nc1ccccc1)c2ccccc2 |
| IUPAC Name | 2-[anilino(phenyl)carbamoyl]hexanoic acid |
| Molecular Weight (g/mol) | 326.39 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?