missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ Buffers for NucleoSpin™ miRNA Plasma Kits
Description
Isolation of small RNA and DNA from plasma and serum
- Efficient and selective isolation of small RNA from plasma or serum samples (patent pending)
- Superior recovery of small RNA ≤ 18 nt
- Optional on-column DNA digest for increased sensitivity in downstream applications
- Simple and fast procedure – without phenol / chloroform
Specifications
Specifications
| Chemical Name or Material | Lysis Buffer MLP |
| CAS | 7646-85-7 |
| Color | Colorless |
| For Use With (Application) | Isolating small RNA from plasma and serum samples |
| For Use With (Equipment) | NucleoSpin™ miRNA Plasma Kit |
| pH | 4.5 to 5.5 |
| Physical Form | Liquid |
| Quantity | 75 mL |
| Formulation | Guanidinium thiocyanate |
| Odor | Odorless |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?