missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Buffers for NucleoSpin™ miRNA Plasma Kits

Product Code. 15859077
Click to view available options
Quantity:
25 mL
75 mL
Chemical Name or Material:
Lysis Buffer MLP
Protein Precipitation Buffer MPP
Unit Size:
25mL
75mL
This item is not returnable. View return policy

Product Code. 15859077

Brand: Macherey-Nagel™ 740365.75

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Supplemental buffer solutions (e.g., lysis buffer, protein precipitation buffer) for use with NucleoSpin™ miRNA Plasma Kits for the rapid isolation of small RNA from from plasma and serum samples.

Isolation of small RNA and DNA from plasma and serum

  • Efficient and selective isolation of small RNA from plasma or serum samples (patent pending)
  • Superior recovery of small RNA ≤ 18 nt
  • Optional on-column DNA digest for increased sensitivity in downstream applications
  • Simple and fast procedure – without phenol / chloroform

Specifications

Chemical Name or Material Lysis Buffer MLP
CAS 7646-85-7
Color Colorless
For Use With (Application) Isolating small RNA from plasma and serum samples
For Use With (Equipment) NucleoSpin™ miRNA Plasma Kit
pH 4.5 to 5.5
Physical Form Liquid
Quantity 75 mL
Formulation Guanidinium thiocyanate
Odor Odorless
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.