missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ Buffer Solutions for NucleoMag™ Virus
Additional buffer solutions (e.g., lysis, binding, wash, carrier RNA) for use in magnetic bead-based isolation of viral RNA and DNA from serum and plasma using the NucleoMag™ Virus kit.
25.40€ - 202.00€
Product Code | Brand | Product Type | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Product Type | Quantity | Price | Quantity & Availability | ||||
15819167
|
Macherey-Nagel™
744872 |
Carrier RNA buffer | 4 x 0.5 mL (2 mL) |
25.40€
2mL |
Estimated Shipment: 12-07-2024
Log in to see stock. |
||||
15830988
|
Macherey-Nagel™
744873.80 |
Lysis buffer MVL | 80 mL |
202.00€
80mL |
Estimated Shipment: 31-05-2024
Log in to see stock. |
||||
15810988
|
Macherey-Nagel™
744867.120 |
Binding buffer MV2 | 250 mL |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
|||||
15809167
|
Macherey-Nagel™
744869.300 |
Wash buffer MV4 | 300 mL |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
|||||
Description
NucleoMag™ Virus Kits
- One-tube procedure minimizes risk of cross-contamination
- Small elution volumes > 50 μL possible
- Yield does not depend on elution volume
- Suitable for manual and automated processing
- Proven 96-well format for high throughput
- Proteinase K and Carrier RNA included
- Extraction of viral DNA and RNA from serum or plasma
- Veterinary testing applications
- Norovirus RNA isolation from stool samples