Learn More
Macherey-Nagel™ Buffer Solutions for NucleoMag™ Virus
Additional buffer solutions (e.g., lysis, binding, wash, carrier RNA) for use in magnetic bead-based isolation of viral RNA and DNA from serum and plasma using the NucleoMag™ Virus kit.
Brand: Macherey-Nagel™ 744872
Additional Details : Weight : 0.00200kg
Description
NucleoMag™ Virus Kits
- One-tube procedure minimizes risk of cross-contamination
- Small elution volumes > 50 μL possible
- Yield does not depend on elution volume
- Suitable for manual and automated processing
- Proven 96-well format for high throughput
- Proteinase K and Carrier RNA included
- Extraction of viral DNA and RNA from serum or plasma
- Veterinary testing applications
- Norovirus RNA isolation from stool samples
Specifications
Colorless | |
5.5 to 6.0 | |
Fusty, moldy | |
Liquid | |
Carrier RNA buffer |
NucleoMag™ Virus kit | |
4 x 0.5 mL (2 mL) | |
Room temperature (18°C to 25 °C) | |
Isolation of viral RNA and DNA from serum or plasma |
Safety and Handling
- carrier RNA buffer
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
- H302-Harmful if swallowed.
- H412-Harmful to aquatic life with long lasting effects.
Supplemental information
- MIXTURE LIST-Contains : guanidinium thiocyanate