missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Buffer Solutions for NucleoMag VET

Product Code. 15870978
Change view
Click to view available options
Quantity:
100 mL
110 mL
Chemical Name or Material:
Binding buffer VEB
Lysis buffer VL1
Unit Size:
100mL
110mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Chemical Name or Material Unit Size
15870978 110 mL Binding buffer VEB 110mL
15869147 100 mL Lysis buffer VL1 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15870978 Supplier Macherey-Nagel™ Supplier No. 744202.110
 Restricted Item
Estimated Shipment: 23-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Additional buffer solutions (e.g., lysis buffer VL1, binding buffer VEB) for use in magnetic bead-based isolation of viral RNA/DNA and bacterial DNA from veterinary samples using the NucleoMag™ VET kit.

NucleoMag™ VET

  • One all-round kit for common veterinary samples
  • High sensitivity even with low viral titers
  • Robust one-tube procedure minimizes risk of cross-contamination
  • Reliable isolation from different viral titers

Specifications

Chemical Name or Material Binding buffer VEB
CAS 64-17-5
Color Colorless
For Use With (Application) Isolation of viral RNA / DNA and bacterial DNA from veterinary samples
For Use With (Equipment) NucleoMag™ VET kit
Physical Form Liquid
Quantity 110 mL
Formulation Sodium perchlorate; Ethanol; Water
Odor Alcoholic
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.