missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Buffer Solutions for NucleoMag 96 Blood

Product Code. 15839157
Change view
Click to view available options
Quantity:
60 mL
100 mL
300 mL
Chemical Name or Material:
Binding Buffer MB2
Elution Buffer MB6
Lysis Buffer MB1
Unit Size:
100mL
300mL
60mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Chemical Name or Material Unit Size
15839157 100 mL Binding Buffer MB2 100mL
15800988 300 mL Elution Buffer MB6 300mL
15890978 60 mL Lysis Buffer MB1 60mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15839157 Supplier Macherey-Nagel™ Supplier No. 744851.80
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Additional buffer solutions (e.g., lysis buffer MB1, binding buffer MB2, elution buffer MB6) for use in magnetic bead-based isolation of pure DNA from whole blood samples using the NucleoMag™ 96 Blood kit.

Specifications

Chemical Name or Material Binding Buffer MB2
CAS 64-17-5
Color Colorless
For Use With (Application) Isolation of DNA from whole blood
For Use With (Equipment) NucleoMag™ 96 Blood
pH 5.0 to 6.0
Physical Form Liquid
Quantity 100 mL
Formulation Ethanol; Sodium perchlorate; Water
Odor Alcoholic
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.